missing translation for 'onlineSavingsMsg'
Learn More

Motic™ Epi-Illuminator, 12V/50W

Código de producto. 15445262
Cambiar vista
Click to view available options
For Use With (Equipment):
BA-310 POL, BA-310 MET Microscopes
Tamaño de la unidad:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. For Use With (Equipment) Tamaño de la unidad
15445262 BA-310 POL, BA-310 MET Microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15445262 Proveedor Motic™ N.º de proveedor 1101001901609
  • ONLINE EXCLUSIVE PRICE
    £1010.00
    £888.88 / Pack of 1
    Save £121.12
    11% Off
Estimated Shipment: 27-01-2027
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Provide illumination for epifluorescence. Motic™ Epi-Illuminator, 12V/50W is a quartz halogen source with integrated field and aperture diaphragms and an external lamp house.

  • Quartz-halogen light source.
  • Integrated field and aperture diaphragms.
  • External lamp house.

Specifications

Type Epi-illumination, Quartz Halogen
Description 12V/50W
For Use With (Equipment) BA-310 POL, BA-310 MET Microscopes
Includes Integrated field and aperture diaphragms and an external lamp house.
Voltage 12 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.