missing translation for 'onlineSavingsMsg'
Learn More

Epi Brassinolide, 90%, TRC

Product Code. 17993015
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17993015 10 mg 10mg
17910955 2.5 mg 2.5mg
17991845 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17993015 Supplier Toronto Research Chemicals Supplier No. TRCE58200010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78821-43-9
Molecular Formula C28 H48 O6
Molecular Weight (g/mol) 480.68
Synonym B-Homo-7-oxaergostan-6-one, 2,3,22,23-tetrahydroxy-, (2α,3α,5α,22R,23R)-, (1R,3aS,3bS,6aS,8S,9R,10aR,10bS,12aS)-1-[(1S,2R,3R,4R)-2,3-Dihydroxy-1,4,5-trimethylhexyl]hexadecahydro-8,9-dihydroxy-10a,12a-dimethyl-6H-benz[c]indeno[5,4-e]oxepin-6-one, (22R,23R,24R)-2α,3α,22,23-Tetrahydroxy-B-homo-7-oxa-5α-ergostan-6-one, 24(R)-Epibrassinolide, 24-Epibrassinolide, 24-epi-Brassinolide, 6H-Benz[c]indeno[5,4-e]oxepin-6-one, 1-(2,3-dihydroxy-1,4,5-trimethylhexyl)hexadecahydro-8,9-dihydroxy-10a,12a-dimethyl-, [1R-[1α(1S*,2R*,3R*,4R*),3aβ,3bα,6aβ,8β,9β,10aα,10bβ,12aα]]-, B 1105, BP 55, Epibrassinolide, Epibrassinolide R, Epin, 6H-Benz[c]indeno[5,4-e]oxepin-6-one, 1-[(1S,2R,3R,4R)-2,3-dihydroxy-1,4,5-trimethylhexyl]hexadecahydro-8,9-dihydroxy-10a,12a-dimethyl-, (1R,3aS,3bS,6aS,8S,9R,10aR,10bS,12aS)-
SMILES CC(C)[C@@H](C)[C@@H](O)[C@H](O)[C@@H](C)[C@H]1CC[C@H]2[C@@H]3COC(=O)[C@H]4C[C@H](O)[C@H](O)C[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C28H48O6/c1-14(2)15(3)24(31)25(32)16(4)18-7-8-19-17-13-34-26(33)21-11-22(29)23(30)12-28(21,6)20(17)9-10-27(18,19)5/h14-25,29-32H,7-13H2,1-6H3/t15-,16+,17+,18-,19+,20+,21-,22+,23-,24-,25-,27-,28-/m1/s1
Formula Weight 480.3451
Chemical Name or Material Epibrassinolide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.