missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPHX4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-84860-0.1ML
This item is not returnable.
View return policy
Description
EPHX4 Polyclonal specifically detects EPHX4 in Human samples. It is validated for Western Blot.
Specifications
| EPHX4 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| ABHD7, abhydrolase domain containing 7, Abhydrolase domain-containing protein 7, EC 3.3, EPHXRPEpoxide hydrolase-related protein, epoxide hydrolase 4, FLJ90341 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human EPHX4 | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 253152 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction