missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EPHX2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143250
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize all reported isoforms (NP_001970.2; NP_001243411.1; NP_001243412.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:2,000.
This gene encodes a member of the epoxide hydrolase family. The protein, found in both the cytosol and peroxisomes, binds to specific epoxides and converts them to the corresponding dihydrodiols. Mutations in this gene have been associated with familial hypercholesterolemia. Alternatively spliced transcript variants have been described.
Specifications
| EPHX2 | |
| Polyclonal | |
| Unconjugated | |
| EPHX2 | |
| bifunctional epoxide hydrolase 2; CEH; cytosolic epoxide hydrolase; Cytosolic epoxide hydrolase 2; Eph2; Ephx2; Epoxide hydratase; epoxide hydrolase 2; epoxide hydrolase 2, cytoplasmic; epoxide hydrolase 2, cytosolic; epoxide hydrolase 2C; epoxide hydrolase, soluble; Lipid-phosphate phosphatase; SEH; sEP; Soluble epoxide hydrolase | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 2053 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P34913 | |
| EPHX2 | |
| Peptide with sequence C-SRTFKSLFRASDES (aa 402-415). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction