missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EPHX2 Monoclonal Antibody (OTI1H5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525223
This item is not returnable.
View return policy
Description
EPHX2 Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the epoxide hydrolase family. The protein, found in both the cytosol and peroxisomes, binds to specific epoxides and converts them to the corresponding dihydrodiols. Mutations in this gene have been associated with familial hypercholesterolemia. Alternatively spliced transcript variants have been described.
Specifications
| EPHX2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P34913 | |
| EPHX2 | |
| Full length human recombinant protein of EPHX2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H5 | |
| Unconjugated | |
| EPHX2 | |
| bifunctional epoxide hydrolase 2; CEH; cytosolic epoxide hydrolase; Cytosolic epoxide hydrolase 2; Eph2; Ephx2; Epoxide hydratase; epoxide hydrolase 2; epoxide hydrolase 2, cytoplasmic; epoxide hydrolase 2, cytosolic; epoxide hydrolase 2C; epoxide hydrolase, soluble; Lipid-phosphate phosphatase; SEH; sEP; Soluble epoxide hydrolase | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2053 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction