missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPHX2 Antibody (S04-2F6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19986-50ul
This item is not returnable.
View return policy
Description
EPHX2 Monoclonal antibody specifically detects EPHX2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| EPHX2 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| S04-2F6 | |
| Unconjugated | |
| CEH, Cytosolic epoxide hydrolase, EC 3.3.2.10, Epoxide hydratase, epoxide hydrolase 2, epoxide hydrolase 2, cytoplasmic, epoxide hydrolase 2, cytosolic, epoxide hydrolase, soluble, SEH, Soluble epoxide hydrolase | |
| A synthetic peptide of human EPHX2 (Uniprot # P34913) | |
| 50 μg | |
| Lipid and Metabolism | |
| 2053 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction