missing translation for 'onlineSavingsMsg'
Learn More

EPHX2 Antibody (OTI1A6), DyLight 594, Novus Biologicals™

Product Code. 18402073 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402073 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402073 Supplier Novus Biologicals Supplier No. NBP270648DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EPHX2 Monoclonal antibody specifically detects EPHX2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPHX2
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1A6
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias CEH, Cytosolic epoxide hydrolase, EC 3.3.2.10, Epoxide hydratase, epoxide hydrolase 2, epoxide hydrolase 2, cytoplasmic, epoxide hydrolase 2, cytosolic, epoxide hydrolase, soluble, SEH, Soluble epoxide hydrolase
Host Species Mouse
Immunogen Full length recombinant protein of human EPHX2 (NP_001970) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2053
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.