missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin-B3 Antibody (MM0264- 12X1), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12283MFV450
This item is not returnable.
View return policy
Description
Ephrin-B3 Monoclonal antibody specifically detects Ephrin-B3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Ephrin-B3 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0264- 12X1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EPH-related receptor tyrosine kinase ligand 8, Ephrin B3, ephrin-B3, EPLG8EPH-related receptor transmembrane ligand ELK-L3, LERK-8, LERK8EFL6 | |
| Human recombinant Ephrin-B3 (extracellular domain) | |
| 0.1 mL | |
| Cardiovascular Biology, Neuroscience | |
| 1949 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction