Learn More
Ephrin B2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26533-1-AP-20UL
 View more versions of this product
Product Code. 18034255
  • £135.00 / 20µL
Description
The Ephrin B2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Ephrin B2. This antibody recognizes human, mouse antigen. The Ephrin B2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
Ephrin B2
400 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Western Blot, ELISA
EFNB2, ephrin B2, EPLG5, Htk L, HTK ligand, HTKL, LERK 5, LERK5
20 μL
1948
Store at -20°C. Stable for one year after shipment.