missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin-B2 Rabbit anti-Human, Mouse, Rat, Clone: 1G9E7, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-15372-100UL
This item is not returnable.
View return policy
Description
Ephrin-B2 Monoclonal antibody specifically detects Ephrin-B2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Ephrin-B2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1G9E7 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| ephrin B2, EPLG5, Htk-L, HTKL, LERK5 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Ephrin-B2 (P52799). | |
| 100 μg | |
| Neuroscience, Signal Transduction | |
| 1948 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction