missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin-B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | Ephrin-B2 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18649261
|
Novus Biologicals
NBP2-98424-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608082
|
Novus Biologicals
NBP2-98424-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ephrin-B2 Polyclonal antibody specifically detects Ephrin-B2 in Rat samples. It is validated for ELISASpecifications
| Ephrin-B2 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 1948 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| ephrin B2, EPLG5, Htk-L, HTKL, LERK5 | |
| Produced in rabbits immunized with purified, recombinant Rat Ephrin-B2 (Accession#: NP_001100798.1; Met1-Ala229). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title