missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Ephrin-B2 Polyclonal antibody specifically detects Ephrin-B2 in Human samples. It is validated for ELISA
Specifications
Specifications
| Conjugate | Janelia Fluor 549 |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Human |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?