missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin-B1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59211
This item is not returnable.
View return policy
Description
Ephrin-B1 Polyclonal antibody specifically detects Ephrin-B1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Ephrin-B | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EFL3, EFL-3, ELK ligand, Elk-L, ephrin-B1, LERK-2 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Bovine: 92%; Rat: 92%. | |
| Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P98172 | |
| EFNB1 | |
| Synthetic peptides corresponding to EFNB1(ephrin-B1) The peptide sequence was selected from the middle region of EFNB1. Peptide sequence SRPSKEADNTVKMATQAPGSRGSLGDSDGKHETVNQEEKSGPGASGGSSG. | |
| 100 μL | |
| Cellular Signaling, Neuroscience | |
| 1947 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction