missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ephrin A4 Polyclonal Antibody

Description
Ephrin A4 Polyclonal Antibody for Western Blot, ICC/IF, IHC, IP
Ephrin A4 is also known as LERK-4 or ELF-4. Ephrin-A4 binds EphA2, EphA3, EphA4, EphA5, EphA6, EphA7, and EphB1. To date, at least 14 members of the Eph receptor family and a family of 8 ligands have been identified. Ligands of Eph family receptors are structurally related membrane-bound proteins that can be subdivided into two major subclasses: ephrin-A and ephrin-B. Ligands in the ephrin-A subclass, including the prototype family member ephrin-A1 (B61), are membrane associated through glycosylphosphatidyl-inositollinkages; whereas ephrin-B subclass consists of ligands with transmembrane domains.
Specifications
Specifications
| Antigen | Ephrin A4 |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene | EFNA4 |
| Gene Accession No. | O08542, P52798 |
| Gene Alias | EFL4; EFL-4; Efna4; EPH-related receptor tyrosine kinase ligand 4; ephrin A4; ephrin-A4; Epl4; EPLG4; LERK4; LERK-4; ligand of eph-related kinase 4 |
| Gene Symbols | EFNA4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?