missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin-A4 Antibody (MM0263-9H13), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12280B
This item is not returnable.
View return policy
Description
Ephrin-A4 Monoclonal specifically detects Ephrin-A4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Ephrin-A4 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| EFNA4 | |
| Human recombinant Ephrin-A4 (extracellular domain) | |
| 0.1 mL | |
| 1945 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0263-9H13 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EFL4, EPH-related receptor tyrosine kinase ligand 4, ephrin-A4, EPLG4MGC125826, LERK-4, LERK4FLJ57652, ligand of eph-related kinase 4 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction