missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | Ephrin A3 |
|---|---|
| Concentration | 0.18 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16870214
|
Proteintech
12480-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16860214
|
Proteintech
12480-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the ephrin (EPH) family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. This gene encodes an EFNA class ephrin.Specifications
| Ephrin A3 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| O08545, P52797 | |
| 13638, 170901, 1944 | |
| Ephrin A3 Fusion Protein Ag3188 | |
| Primary | |
| -20°C | |
| EFNA3 |
| 0.18 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| EFL 2, EFL2, EFNA3, Ehk1 L, EHK1 ligand, ephrin A3, EPLG3, LERK 3, LERK3 | |
| Efna3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title