missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin-A3 Antibody (MM0262-7N12), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12279AF647
This item is not returnable.
View return policy
Description
Ephrin-A3 Monoclonal specifically detects Ephrin-A3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Ephrin-A3 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EFNA3 | |
| Human recombinant Ephrin-A3 (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0262-7N12 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EFL2, EFL-2, EHK1-L, EPH-related receptor tyrosine kinase ligand 3, ephrin-A3, EPLG3EHK1 ligand, LERK3LERK-3, ligand of eph-related kinase 3 | |
| Mouse | |
| Protein G purified | |
| Growth and Development, Neuronal Cell Markers | |
| 1944 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction