missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin-A2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98103CL1
This item is not returnable.
View return policy
Description
Ephrin-A2 Polyclonal antibody specifically detects Ephrin-A2 in Mouse samples. It is validated for ELISA
Specifications
| Ephrin-A2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 1943 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| CoraFluor 1 | |
| ELF-1, EPH-related receptor tyrosine kinase ligand 6, ephrin-A2, EPLG6HEK7 ligand, HEK7-L, HEK7-ligand, LERK6LERK-6, ligand of eph-related kinase 6 | |
| Produced in rabbits immunized with purified, recombinant Mouse Ephrin-A2 (Accession#: NP_031935.3; Met 1-Asn 184) | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction