Learn More
Invitrogen™ Ephrin A1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ 343300
Product Code. 10602964
  • £444.00 / 100µg
For Research Use Only. All usage must comply with product instructions.
Description
Description
Ephrin A1 Polyclonal Antibody for Western Blot, ICC/IF, IHC, IP
This gene encodes a member of the ephrin family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B class, which are transmembrane proteins. This gene encodes an EFNA class ephrin which binds to the EPHA2, EPHA4, EPHA5, EPHA6, and EPHA7 receptors. Two transcript variants that encode different isoforms were identified through sequence analysis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ephrin A1
Polyclonal
Unconjugated
EFNA1
AI325262; B61; ECKLG; EFL1; Efna1; Epgl1; EPH-related receptor tyrosine kinase ligand 1; ephrin A1; ephrin-A1; Ephrin-A1, secreted form; Epl1; EPLG1; Immediate early response protein B61; Lerk1; LERK-1; ligand of eph-related kinase 1; TNF alpha-induced protein 4; TNFAIP4; tumor necrosis factor alpha-induced protein 4; tumor necrosis factor, alpha-induced protein 4
Rabbit
Antigen Affinity Chromatography
RUO
13636, 1942
-20°C
Liquid
Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
0.25 mg/mL
PBS with 0.1% sodium azide; pH 7.4
P20827, P52793
EFNA1
Synthetic peptide derived from the C-terminal end of mouse ephrin-A1 protein.
100 μg
Primary
Human, Mouse