missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Ephrin-A1 Antibody Pair [HRP]

Product Code. p-7183252 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18202537

Brand: Novus Biologicals™ NBP27931215PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Ephrin-A1.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 39 pg/mL
Assay Sensitivity 39.06-2500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Ephrin-A1.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4
Gene ID (Entrez) 1942
Gene Symbol EFNA1
Host Species Rabbit/Mouse
Kit Contents
  • Standard: Each vial contains 140 ng of recombinant Ephrin-A1.
  • Capture Antibody: 0.5 mg/mL of mouse anti-Ephrin-A1 monoclonal antibody,
  • Detection Antibody: 1 mg/mL rabbit anti-Ephrin-A1 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer
    Storage Requirements Storage is content dependent.
    Target Ephrin-A1
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.2 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 2500 pg/mL is recommended.
    Vis mere Vis mindre
    Korrektion af produktindhold

    Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

    Produkttitel

    Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

    Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.