missing translation for 'onlineSavingsMsg'
Learn More

Ephrin-A1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NBP1-52103

Product Code. 18231245

  • £407.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ephrin-A1 Polyclonal specifically detects Ephrin-A1 in Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ephrin-A1
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4
Goat
Affinity Purified
RUO
Primary
This antibody is expected to recognize isoform a (NP_004419.2) only.
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, ELISA
0.5 mg/mL
Western Blot 0.01 - 0.03 ug/ml, Peptide ELISA Detection limit 1:16000
NP_004419.2
EFNA1
Peptide with sequence C-KVTVSGKITHSP corresponding to internal region according to NP_004419.2.
0.1 mg
Cancer
1942
Mouse, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only