missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin-A1 Antibody (CL6501) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76497AF750
This item is not returnable.
View return policy
Description
Ephrin-A1 Monoclonal antibody specifically detects Ephrin-A1 in Human samples. It is validated for Western Blot
Specifications
| Ephrin-A1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4 | |
| This antibody was developed using a recombinant protein derived from P20827, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 1942 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL6501 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction