missing translation for 'onlineSavingsMsg'
Learn More

Ephrin-A1 Antibody (067), DyLight 650 , Novus Biologicals™

Product Code. 18411878 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411878 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411878 Supplier Novus Biologicals Supplier No. NBP289659C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Ephrin-A1 Monoclonal antibody specifically detects Ephrin-A1 in Rat samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ephrin-A1
Applications ELISA
Classification Monoclonal
Clone 067
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Rat Ephrin-A1 (P97553; Met1-His181).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1942
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.