missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Antibody detects endogenous levels of total EPHB6.
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The ephrin receptor encoded by this gene lacks the kinase activity of most receptor tyrosine kinases and binds to ephrin-B ligands.
Specifications
Specifications
| Antigen | EphB6 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | EPHB6 |
| Gene Accession No. | O08644, O15197, P0C0K7 |
| Gene Alias | Cekl; chicken eph/elk receptor-like protein; EPB6; EPH receptor B6; EPHB6; EphB6v; Epherin B6; Epherin receptor B6; ephrin type A receptor; ephrin type-B receptor 6; HEP; human kinase-defective Eph-family receptor protein; MEP; MGC129910; MGC129911; Tyrosine-protein kinase-defective receptor EPH-6 |
| Gene Symbols | EPHB6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
