missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB6 Antibody (465327) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB33841JF669
This item is not returnable.
View return policy
Description
EphB6 Monoclonal antibody specifically detects EphB6 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| EphB6 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, HEPTyrosine-protein kinase-defective receptor EPH-6, MGC129910, MGC129911 | |
| Mouse myeloma cell line NS0-derived recombinant human EphB6, Val17-Thr579, Accession # O15197 | |
| 0.1 mL | |
| Protein Kinase | |
| 2051 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 465327 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction