missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EphB4 Recombinant Rabbit Monoclonal Antibody (23GB1045), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | EphB4 |
|---|---|
| Clone | 23GB1045 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088802
![]() |
Invitrogen™
MA554056 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088480
|
Invitrogen™
MA554057 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EphB4 Recombinant Monoclonal antibody specifically detects EphB4 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| EphB4 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P54760, P54761 | |
| 13846, 2050, 686310 | |
| IgG | |
| Affinity Chromatography | |
| Antibody |
| 23GB1045 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI042935; Developmental kinase 2; EPH receptor B4; Ephb4; ephrin receptor EphB4; ephrin type-B receptor 4; Hepatoma transmembrane kinase; HTK; Mdk2; mDK-2; MYK1; soluble EPHB4 variant 1; soluble EPHB4 variant 2; soluble EPHB4 variant 3; TYRO11; Tyrosine kinase MYK-1; tyrosine-protein kinase receptor HTK; tyrosine-protein kinase TYRO11 | |
| A synthetic peptide of human Eph receptor B4/HTK (236-529AA). | |
| Primary | |
| -20°C | |
| EPHB4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title