Learn More
Eurofins DiscoverX EphB4 Protein, Active
Description
Eph family tyrosine kinase receptors consist of two groups: EphA group members respond to ephrin-A, which is anchored to the cell membrane by glycosylphosphatidylinositol, whereas EphB group members respond to ephrin-B, containing a transmembrane domain. Among them, only EphA4 can cross-respond to both ephrin-A and -B. Expression of the EPB4 (EphB4, Ephrin type-B receptor 4, Tyrosine-protein kinase receptor HTK, EC 2.7.1.112) receptors and their ephrinB ligands play a key role in normal vascular formation as well as in diabetic retinopathy, malignant tumor progression and angiogenesis. Inhibition of EPB4 may be used in the treatment of diabetic renopathy and angioproliferative tumors.
GenBank AY056047
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 51 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥73% |
| Protein | EphB4, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.