Learn More
EphB4 Monoclonal Antibody (5B8F7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515286
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-15286 targets EphB4 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15286 immunogen is purified recombinant fragment of EphB4 expressed in E. Coli.
EphB4 Monoclonal antibody specifically detects EphB4 in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
EphB4 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P54760 | |
EPHB4 | |
Purified recombinant fragment of EphB4 expressed in E. Coli. | |
RUO | |
2050 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
5B8F7 | |
Unconjugated | |
EPHB4 | |
AI042935; Developmental kinase 2; EPH receptor B4; Ephb4; ephrin receptor EphB4; ephrin type-B receptor 4; Hepatoma transmembrane kinase; HTK; Mdk2; mDK-2; MYK1; soluble EPHB4 variant 1; soluble EPHB4 variant 2; soluble EPHB4 variant 3; TYRO11; Tyrosine kinase MYK-1; tyrosine-protein kinase receptor HTK; tyrosine-protein kinase TYRO11 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |