missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB4 Antibody (VEB4-7E4), Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-49607-0.05MG
This item is not returnable.
View return policy
Description
EphB4 Monoclonal specifically detects EphB4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoassay.
Specifications
| EphB4 | |
| Monoclonal | |
| 0.1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay | |
| EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, HTKephrin receptor EphB4, MYK1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse Ephrin Type-B Receptor 4 | |
| Store at 4C. Do not freeze. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| VEB4-7E4 | |
| Unconjugated | |
| PBS, containing 0.1% BSA with 0.02% Sodium Azide | |
| EPHB4 | |
| Mouse Tyrosine-protein kinase receptor HTK | |
| 0.05 mg | |
| Protein Kinase | |
| 2050 | |
| Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction