missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ EphB4 Antibody (Morphosys patent anti-EphB4) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28530-100ug
This item is not returnable.
View return policy
Description
EphB4 Monoclonal antibody specifically detects EphB4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| EphB4 | |
| Monoclonal | |
| Unconjugated | |
| P54760 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Morphosys patent anti-EphB4 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, HTKephrin receptor EphB4, MYK1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11 | |
| EphB4 | |
| 100 μg | |
| Protein Kinase | |
| 2050 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction