missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB3 Rabbit anti-Mouse, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90644-100UL
This item is not returnable.
View return policy
Description
EphB3 Monoclonal specifically detects EphB3 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| EphB3 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 1 | |
| Flow Cytometry 1:25-1:100 | |
| EC 2.7.10, EC 2.7.10.1, EK2, EPH receptor B3, EphB3, EPH-like kinase 2, EPH-like tyrosine kinase-2, ephrin type-B receptor 3, ETK2Hek2, HEK2, human embryo kinase 2, Tyro6, Tyrosine-protein kinase TYRO6 | |
| Recombinant Mouse EphB3 Protein | |
| 100 μg | |
| Protein Kinase | |
| 2049 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction