missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB3 Antibody (647354) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB56671JF585
This item is not returnable.
View return policy
Description
EphB3 Monoclonal antibody specifically detects EphB3 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| EphB3 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| EC 2.7.10, EC 2.7.10.1, EK2, EPH receptor B3, EphB3, EPH-like kinase 2, EPH-like tyrosine kinase-2, ephrin type-B receptor 3, ETK2Hek2, HEK2, human embryo kinase 2, Tyro6, Tyrosine-protein kinase TYRO6 | |
| Mouse myeloma cell line NS0-derived recombinant human EphB3, Leu38-Ala550, Accession # P54753 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 647354 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Protein Kinase | |
| 2049 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction