missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EphB3 Monoclonal antibody specifically detects EphB3 in Mouse samples. It is validated for ELISA
Specifications
Specifications
| Gene Alias | EC 2.7.10, EC 2.7.10.1, EK2, EPH receptor B3, EphB3, EPH-like kinase 2, EPH-like tyrosine kinase-2, ephrin type-B receptor 3, ETK2Hek2, HEK2, human embryo kinase 2, Tyro6, Tyrosine-protein kinase TYRO6 |
| Host Species | Rabbit |
| Regulatory Status | RUO |
| Target Species | Mouse |
| Content And Storage | Store at 4C in the dark. |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?