missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB2 Antibody (Genentech patent anti-EphB2) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28529JF585
This item is not returnable.
View return policy
Description
EphB2 Monoclonal antibody specifically detects EphB2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| EphB2 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CAPB, DRTEphB2, EC 2.7.10, EC 2.7.10.1, EK5, elk-related tyrosine kinase, EPH receptor B2, eph tyrosine kinase 3, EPH-like kinase 5, ephrin type-B receptor 2, EPHT3MGC87492, EPTH3, ERKHek5, HEK5, PCBC, protein-tyrosine kinase HEK5, Renal carcinoma antigen NY-REN-47, Tyro5, Tyrosine-protein kinase receptor EPH-3, Tyrosine-protein kinase TYRO5 | |
| EphB2 | |
| 0.1 mL | |
| Prostate Cancer, Protein Kinase | |
| 2048 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Genentech patent anti-EphB2 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction