missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB2 Antibody (2D12C6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47525
This item is not returnable.
View return policy
Description
EphB2 Monoclonal antibody specifically detects EphB2 in Human, Porcine samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications
| EphB2 | |
| Monoclonal | |
| Unconjugated | |
| CAPB, DRTEphB2, EC 2.7.10, EC 2.7.10.1, EK5, elk-related tyrosine kinase, EPH receptor B2, eph tyrosine kinase 3, EPH-like kinase 5, ephrin type-B receptor 2, EPHT3MGC87492, EPTH3, ERKHek5, HEK5, PCBC, protein-tyrosine kinase HEK5, Renal carcinoma antigen NY-REN-47, Tyro5, Tyrosine-protein kinase receptor EPH-3, Tyrosine-protein kinase TYRO5 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Pig | |
| Ascites |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 2D12C6 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| EPHB2 | |
| Purified recombinant fragment of EphB2(aa17-200) expressed in E. Coli. | |
| 0.1 mL | |
| Prostate Cancer, Protein Kinase | |
| 2048 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction