missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB1 Rabbit anti-Human, Clone: 22, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90032-100UL
This item is not returnable.
View return policy
Description
EphB1 Monoclonal specifically detects EphB1 in Human samples. It is validated for ELISA.
Specifications
| EphB1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 22 | |
| ELISA 1:5000-1:10000 | |
| EC 2.7.10, EC 2.7.10.1, EK6, ELK, EPH receptor B1, eph tyrosine kinase 2, EphB1, EPH-like kinase 6, ephrin type-B receptor 1, EPHT2FLJ37986, hEK6, NETHek6, soluble EPHB1 variant 1, Tyrosine-protein kinase receptor EPH-2 | |
| Recombinant Human EphB1 protein | |
| 100 μg | |
| Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase | |
| 2047 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction