missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB1/EphB2 Polyclonal Antibody, Invitrogen™
Description
The Eph subfamily represents the largest group of receptor protein tyrosine kinases identified to date. While the biological activities of these receptors have yet to be determined, there is increasing evidence that they are involved in central nervous system function and in development. The Eph subfamily receptors of human origin (and their murine/avian homologs) include EphA1(Eph), EphA2 (Eck), EphA3 (Hek4), EphA4 (Hek8), EphA5 (Hek7), EphA6 (Hek12),EphA7 (Hek11/MDK1), EphA8 (Hek3), EphB1 (Hek6), EphB2 (Hek5), EphB3(Cek10, Hek2), EphB4 (Htk), EphB5 (Hek9) and EphB6 (Mep). Ligands for Eph receptors include ephrin-A4 (LERK-4) which binds EphA3 and EphB1. In addition, Ephrin-A2 (Elf-1) has been described as the ligand for EphA4, ephrin-A3 (Ehk1-L) as the ligand for EphA5 and ephrin-B2 (Htk-L) as the ligand for EphB4 (Htk).
Specifications
Specifications
| Antigen | EphB1/EphB2 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | EPHB1 |
| Gene Accession No. | P29323, P54762, P54763, Q8CBF3, P09759 |
| Gene Alias | ETK2; HEK2; TYRO6; EPHB3, DRT; ERK; CAPB; Hek5; PCBC; EPHT3; Tyro5; MGC87492; EPHB2, Ephrin type-B receptor 1; Tyrosine-protein kinase receptor EPH-2; NET, HEK-6, ELK, Ephrin type-B receptor 2, Ephrin B1, EphrinB1, Ephrin B2, EphrinB2 |
| Gene Symbols | EPHB1, EPHB2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?