missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB1 Antibody (5F10A4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28887-0.025ml
This item is not returnable.
View return policy
Description
EphB1 Monoclonal specifically detects EphB1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| EphB1 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| EC 2.7.10, EC 2.7.10.1, EK6, ELK, EPH receptor B1, eph tyrosine kinase 2, EphB1, EPH-like kinase 6, ephrin type-B receptor 1, EPHT2FLJ37986, hEK6, NETHek6, soluble EPHB1 variant 1, Tyrosine-protein kinase receptor EPH-2 | |
| Mouse | |
| 110 kDa | |
| 0.025 mL | |
| Growth and Development, Neuronal Cell Markers, Neuroscience, Protein Kinase | |
| 2047 | |
| Human | |
| Ascites |
| Western Blot | |
| 5F10A4 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| P54762 | |
| EPHB1 | |
| Purified recombinant fragment of EPH Receptor B1(aa19-133) expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Specific for EPH Receptor B1. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction