missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB1 Antibody (022), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90032IR
This item is not returnable.
View return policy
Description
EphB1 Monoclonal specifically detects EphB1 in Human samples. It is validated for ELISA.
Specifications
| EphB1 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human EPHB4 Human EPHB6 Mouse EphA1 Mouse EphA4 Mouse EphA6 Mouse EphA7 Mouse EphB3 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 22 | |
| ELISA | |
| EC 2.7.10, EC 2.7.10.1, EK6, ELK, EPH receptor B1, eph tyrosine kinase 2, EphB1, EPH-like kinase 6, ephrin type-B receptor 1, EPHT2FLJ37986, hEK6, NETHek6, soluble EPHB1 variant 1, Tyrosine-protein kinase receptor EPH-2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human EphB1 (P54762-1; Met1-Pro540). | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase | |
| 2047 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction