missing translation for 'onlineSavingsMsg'
Learn More

EphA8 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18365975 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365975 100 μL 100µL
18325126 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18365975 Supplier Novus Biologicals Supplier No. NBP304924100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EphA8 Polyclonal specifically detects EphA8 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias EEK, EK3, EPH- and ELK-related kinase, EPH- and ELK-related tyrosine kinase, EPH receptor A8, EPH-like kinase 3, ephrin type-A receptor 8, HEK3, hydroxyaryl-protein kinase, KIAA1459, protein-tyrosine kinase, tyrosine-protein kinase receptor EEK, tyrosylprotein kinase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 28-300 of human EPHA8 (NP_001006944.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2046
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.