Learn More
Eurofins DiscoverX EphA8 Protein, Active
Description
Eph (erythropoietin-producing hepatocellular) family tyrosine kinase receptors consist of two groups: EphA group members respond to ephrin-A, which is anchored to the cell membrane by glycosylphosphatidylinositol, whereas EphB group members respond to ephrin-B, containing a transmembrane domain. Among them, only EphA4 can cross-respond to both ephrin-A and -B. The expression of EphA8 receptor significantly promoted cell attachment to fibronectin without altering cellular integrin levels. Moreover, p110 phosphotidylinositol (PI) 3-kinase plays as a key bridging molecule between EphA8 and integrins. Inhibition. of EphA8 may disrupt establishment, maintenance, and remodeling of patterns of cellular organization and may also be used in the treatment of various cancers.
GenBank NM_020526
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 37.1 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥64% |
| Protein | EphA8, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.