missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EphA7, GST Tag Recombinant Protein

Product Code. 15418513
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
This item is not returnable. View return policy

Product Code. 15418513

Brand: Invitrogen™ PR7046A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic

EPHA7 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA7 is involved in embryonic development. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15375
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 2045
Molecular Weight (g/mol) 76.7 kDa
Name Human EphA7, GST Tag
Quantity 100 μg
Regulatory Status RUO - research use only
Gene Alias Cek11; Developmental kinase 1; Ebk; EHK3; EHK-3; EK11; embryonic brain kinase; epa7; EPH homology kinase 3; Eph homology kinase-3; EPH receptor A7; Epha7; EPHA7-S; EPH-like kinase 11; ephrin receptor EphA7; ephrin type-A receptor 7; HEK11; Mdk1; mDK-1; receptor protein-tyrosine kinase HEK11; tyrosine-protein kinase receptor EHK-3
Common Name EphA7
KinaseFamily EPH Receptor Kinase Family
Protein Length 579-998
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.