Learn More
Invitrogen™ Human EphA7, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic
Specifications
Specifications
| Accession Number | Q15375 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 2045 |
| Molecular Weight (g/mol) | 76.7 kDa |
| Name | Human EphA7, GST Tag |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Gene Alias | Cek11; Developmental kinase 1; Ebk; EHK3; EHK-3; EK11; embryonic brain kinase; epa7; EPH homology kinase 3; Eph homology kinase-3; EPH receptor A7; Epha7; EPHA7-S; EPH-like kinase 11; ephrin receptor EphA7; ephrin type-A receptor 7; HEK11; Mdk1; mDK-1; receptor protein-tyrosine kinase HEK11; tyrosine-protein kinase receptor EHK-3 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.