missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA7 Antibody (6E1), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB110-93606G
This item is not returnable.
View return policy
Description
EphA7 Monoclonal specifically detects EphA7 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| EphA7 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EPHA7 | |
| Mouse EphA7 (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| This was selected for its ability to detect mouse EphA7 in western blots. In this format, this does not cross-react with recombinant mouse EphA2, EphA3, EphA4, EphA6, EphA8 and EphB2, EphB3 or EphB4. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 60 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3 | |
| Rat | |
| Protein A or G purified | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Kinase | |
| 2045 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction