Learn More
Invitrogen™ Human EphA6, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant human EPHA6 (561-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant The gene accession number for EphA6 is NP_001073917.2.
Specifications
Specifications
| Accession Number | Q9UF33 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 285220 |
| Molecular Weight (g/mol) | 77 kDa |
| Name | Human EphA6, GST Tag |
| Quantity | 10 μg |
| Gene Alias | DKFZp434C1418; EHK2; EHK-2; EK12; EPA6; EPH homology kinase 2; Eph receptor A6; EPHA6; EPH-like kinase 12; ephrin receptor EphA6; ephrin type-A receptor 6; FLJ35246; HEK12; m-ehk2; PRO57066; tyrosine-protein kinase receptor EHK-2 |
| Common Name | EphA6 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.