Learn More
Invitrogen™ Human EphA5, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic
Specifications
Specifications
| Accession Number | P54756 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris HCl with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 2044 |
| Molecular Weight (g/mol) | 77.1 kDa |
| Name | Human EphA5, GST Tag |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Gene Alias | AI854630; ARCC2; AW125296; brain-specific kinase; BSK; CEK7; CEK-7; CTPA; CTPP1; ECK; eck-like sequence 1; EHK1; EHK-1; EK7; Ekh1; Els1; Els1.; epa5; EPH homology kinase 1; Eph receptor A5; EPHA5; EPH-like kinase 7; Ephrin A2; Ephrin A5; Ephrin type-A receptor 5; EphrinA2; EphrinA5; HEK7; receptor protein-tyrosine kinase HEK7; Rek7; TYRO4; tyrosine-protein kinase receptor EHK-1 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.