missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EphA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA514578
This item is not returnable.
View return policy
Description
This antibody is predicted to react with chicken, mouse and Xenopus based on sequence homology.
EPHA4 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EphA4 encodes a protein that binds ephrin-A ligands.
Specifications
| EphA4 | |
| Polyclonal | |
| Unconjugated | |
| EPHA4 | |
| 2900005C20Rik; AI385584; Cek8; Cek8 protein; EK8; epa4; EPH receptor A4; Epha4; EPH-like kinase 8; Ephrin type-A receptor 4; Hek8; rb; receptor protein-tyrosine kinase HEK8; RGD1560587; Sek; Sek1; Tyro1; TYRO1 protein tyrosine kinase; tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor SEK; tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase TYRO1 | |
| Rabbit | |
| Size-exclusion - Dialysis, Ammonium sulfate precipitation | |
| RUO | |
| 2043 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P54764 | |
| EPHA4 | |
| KLH conjugated synthetic peptide between 40-70 amino acids from the N-terminal region of human EphA4. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering