missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EphA4 Monoclonal Antibody (M280)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547715
This item is not returnable.
View return policy
Description
This antibody detects 95 and 120 kDa* bands corresponding to EphA4 in Western blots of HUVEC cells and a 120 kDa band in mouse brain tissue.
EPHA4 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EphA4 encodes a protein that binds ephrin-A ligands.
Specifications
| EphA4 | |
| Monoclonal | |
| 0.25 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P54764, Q03137 | |
| EPHA4 | |
| Clone M280 was generated from a recombinant protein corresponding to amino acids in the N-terminal region of human EphA4. This region has less than 50% homology to similar regions in other EphA and EphB family members. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| M280 | |
| Unconjugated | |
| EPHA4 | |
| 2900005C20Rik; AI385584; Cek8; Cek8 protein; EK8; epa4; EPH receptor A4; Epha4; EPH-like kinase 8; Ephrin type-A receptor 4; Hek8; rb; receptor protein-tyrosine kinase HEK8; RGD1560587; Sek; Sek1; Tyro1; TYRO1 protein tyrosine kinase; tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor SEK; tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase TYRO1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 13838, 2043, 316539 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction