missing translation for 'onlineSavingsMsg'
Learn More

EphA4 Antibody (6H7), Novus Biologicals™

Product Code. 18359737 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359737 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359737 Supplier Novus Biologicals Supplier No. H00002043M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EphA4 Monoclonal antibody specifically detects EphA4 in Human, Mouse samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA4
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 6H7
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA 1:100-1:2000
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004429
Gene Alias EC 2.7.10, EC 2.7.10.1, EK8, EPH receptor A4, EphA4, EPH-like kinase 8, ephrin type-A receptor 4, HEK8, receptor protein-tyrosine kinase HEK8, SEK, TYRO1, TYRO1 protein tyrosine kinase, Tyrosine-protein kinase receptor SEK, Tyrosine-protein kinase TYRO1
Host Species Mouse
Immunogen EPHA4 (NP_004429, 887 a.a. ~ 986 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SLKRTGTESSRPNTALLDPSSPEFSAVVSVGDWLQAIKMDRYKDNFTAAGYTTLEAVVHVNQEDLARIGITAITHQNKILSSVQAMRTQMQQMHGRMVPV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2043
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.