Learn More
Invitrogen™ EphA2 Monoclonal Antibody (M049)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547665
Description
Clone M049 mouse monoclonal antibody detects a 125kDa* protein on SDS-PAGE Native immunoblots of human NCI-H2052 and MDA-MB-231 carcinomas, as well as the EphA2 C-terminal 60 kDa fragment in human A431 and A549. The antibody detects native EphA2, as well as aldehyde-fixed EphA2. The antibody works for Native western blot, immunoprecipitation, immunocytochemistry, and ELISA capture.
EPHA2 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA2 is overexpressed in many types of human cancers. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e.g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e.g. ABL, FAK, JAK, and SRC families).
Specifications
| EphA2 | |
| Monoclonal | |
| 0.25 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P29317 | |
| EPHA2 | |
| Clone (M049) was generated from a recombinant protein that included the extracellular region of human EphA2 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| M049 | |
| Unconjugated | |
| EPHA2 | |
| ARCC2; AW545284; CTPA; CTPP1; CTRCT6; Eck; EPH receptor A2; Epha2; ephrin receptor EphA2; Ephrin type-A receptor 2; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; I79_019726; Myk2; Sek2; Sek-2; soluble EPHA2 variant 1; tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor MPK-5; tyrosine-protein kinase receptor SEK-2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 1969 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.